11319
4,4″-Bis(4,4,4-trifluoro-1,3-dioxobutyl)-o-terphenyl-4′-sulfonyl chloride
BioReagent, suitable for fluorescence, ≥95.0% (HPCE)
Synonym(s):
1,1′-[4′-(Chlorosulfonyl)-o-terphenyl-4,4″-diyl]bis(4,4,4-trifluoro-1,3-butanedione)
About This Item
product line
BioReagent
assay
≥95.0% (HPCE)
fluorescence
λex 385 nm; λem 614 nm in 0.1 M Tris pH 8.0, Eu3+
suitability
suitable for fluorescence
SMILES string
FC(F)(F)C(=O)CC(=O)c1ccc(cc1)-c2ccc(cc2-c3ccc(cc3)C(=O)CC(=O)C(F)(F)F)S(Cl)(=O)=O
InChI
1S/C26H15ClF6O6S/c27-40(38,39)18-9-10-19(14-1-5-16(6-2-14)21(34)12-23(36)25(28,29)30)20(11-18)15-3-7-17(8-4-15)22(35)13-24(37)26(31,32)33/h1-11H,12-13H2
InChI key
CJRMAUPUOUJYMF-UHFFFAOYSA-N
Application
Packaging
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service