Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10118

Sigma-Aldrich

Amylopectin from potato starch

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201

impurities

≤10% water

InChI

1S/C30H52O26/c31-1-6-11(35)13(37)19(43)28(50-6)55-24-9(4-34)52-27(21(45)16(24)40)48-5-10-25(56-29-20(44)14(38)12(36)7(2-32)51-29)17(41)22(46)30(53-10)54-23-8(3-33)49-26(47)18(42)15(23)39/h6-47H,1-5H2/t6-,7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26+,27+,28-,29-,30-/m1/s1

Inchi Key

WMGFVAGNIYUEEP-WUYNJSITSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Branching enzyme assay

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C R Krisman et al.
Analytical biochemistry, 147(2), 491-496 (1985-06-01)
Methods previously described for glycogen or amylopectin branching enzymatic activity are insufficiently sensitive and not quantitative. A new, more sensitive, specific, and quantitative one was developed. It is based upon the quantitation of the glucose residues joined by alpha 1,6
Varatharajan Vamadevan et al.
Carbohydrate polymers, 92(2), 1653-1659 (2013-02-13)
The relationship between the internal structure of amylopectin from diverse plants and thermal properties of the starch granules has been investigated. Correlations were found between structural parameters, such as number of building blocks in clusters, interblock chain length and length
Li Guo et al.
Carbohydrate polymers, 92(1), 441-447 (2012-12-12)
The compatibilities of amylose/tea polysaccharide (Am/TPS) and amylopectin/tea polysaccharide (Ap/TPS) in water were investigated with theoretical calculations and experimental measurements. To achieve this, dilute-solution viscometry (DSV) and high-speed differential scanning calorimetry (hyper-DSC) were used. The compatibility criteria on the basis
Anna Källman et al.
International journal of biological macromolecules, 55, 75-82 (2012-12-29)
Amylopectin is a highly branched starch component built up of a large number of clustered α-D-glucose chains. A single C-chain possesses the reducing end and carries the rest of the macromolecule. The aim of this study was to investigate the
Masahiro Murase et al.
ChemSusChem, 5(12), 2307-2311 (2012-11-22)
Rice to power: Amylopectin is a major component of agricultural products such as corn, potato, and rice. Silicon-graphite electrodes are prepared by using slurries of these polysaccharides as binders. Compared to the conventionally used binder PVdF, they exhibit drastically improved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service