Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08747

Sigma-Aldrich

Chromeo P465

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

fluorescence

λex 486 nm; λem 650 nm±10 nm (in buffer pH 9, BSA)

storage temp.

−20°C

General description

Chromeo P465 labels proteins and peptides by exhibiting a color change from blue to red upon binding to primary amines. Chromeo P465 displays a weak fluorescence with a quantum yield <1% in solution. After conjugation to a primary amine group, the label undergoes a shortwave spectral shift of >100 nm and the quantum yield rises to 14%. This property allows a distinct detection of primary amines, proteins, and other biomolecules.

Caution

To ensure stability, the lyophilized dye should be stored at 4°C in the dark. This product is guaranteed for 6 months from the date of arrival.

Legal Information

Chromeo is a trademark of Active Motif Chromeon GmbH

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service