Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08187

Sigma-Aldrich

L-Sorbose-1-phosphate lithium salt

≥90% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O9P · xLi+
Molecular Weight:
259.13 (free acid basis)
UNSPSC Code:
12352201
NACRES:
NA.25

assay

≥90% (TLC)

storage temp.

−20°C

Biochem/physiol Actions

Substrate of L-sorbose-1-phosphate reductase[1][2]; strongly inhibits brain and tumor hexokinase[3].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The mechanism by which glyceraldehyde inhibits glycolysis.
H A LARDY et al.
The Journal of biological chemistry, 187(1), 325-337 (1950-11-01)
L-Sorbose-1-phosphate reductase.
R L Anderson et al.
Methods in enzymology, 89 Pt D, 248-251 (1982-01-01)
Wichai Soemphol et al.
Journal of bacteriology, 189(13), 4800-4808 (2007-05-01)
Upstream of the gene for flavin adenine dinucleotide (FAD)-dependent D-sorbitol dehydrogenase (SLDH), sldSLC, a putative transcriptional regulator was found in Gluconobacter frateurii THD32 (NBRC 101656). In this study, the whole sboR gene and the adjacent gene, sboA, were cloned and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service