Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07859

Sigma-Aldrich

SARS Protease Substrate

≥85% (HPLC)

Synonym(s):

L-Valyl-L-seryl-L-valyl-L-asparaginyl-L-seryl-L-threonyl-L-leucyl-L-glutaminyl-L-serylglycyl-L-leucyl-L-arginyl-L-lysyl-L-methionyl-L-alanine, Val-Ser-Val-Asn- Ser-Thr-Leu-Gln-Ser-Gly-Leu-Arg-Lys-Met-Ala

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H119N21O22S
CAS Number:
Molecular Weight:
1590.84
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

assay

≥85% (HPLC)

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](CO)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@H](CO)NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](NC(=O)[C@H](CO)NC(=O)[C@@H](N)C(C)C)C(C)C)[C@@H](C)O)C(=O)N[C@@H](C)C(O)=O

InChI

1S/C66H119N21O22S/c1-30(2)23-40(57(100)78-37(16-14-21-73-66(71)72)55(98)77-36(15-12-13-20-67)54(97)80-39(19-22-110-11)53(96)75-34(9)65(108)109)76-48(94)26-74-52(95)43(27-88)83-56(99)38(17-18-46(68)92)79-58(101)41(24-31(3)4)81-64(107)51(35(10)91)87-61(104)44(28-89)84-59(102)42(25-47(69)93)82-63(106)50(33(7)8)86-60(103)45(29-90)85-62(105)49(70)32(5)6/h30-45,49-51,88-91H,12-29,67,70H2,1-11H3,(H2,68,92)(H2,69,93)(H,74,95)(H,75,96)(H,76,94)(H,77,98)(H,78,100)(H,79,101)(H,80,97)(H,81,107)(H,82,106)(H,83,99)(H,84,102)(H,85,105)(H,86,103)(H,87,104)(H,108,109)(H4,71,72,73)/t34-,35+,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,49-,50-,51-/m0/s1

InChI key

KPAXWJVUQQAPFD-KZVXTNNVSA-N

Application

Enzyme substrate for the assays of the SARS coronavirus main protease[1]

Biochem/physiol Actions

A study for determining substrate specificity for the SARS main protease indicated that the best substrates contained residues with high β-sheet propensity at positions P5 and P3., small hydrophobic residues at P4, hydrophobic residues without β-branching at residue P2 and a GLN for residue P1. [2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kanchan Anand et al.
Science (New York, N.Y.), 300(5626), 1763-1767 (2003-05-15)
A novel coronavirus has been identified as the causative agent of severe acute respiratory syndrome (SARS). The viral main proteinase (Mpro, also called 3CLpro), which controls the activities of the coronavirus replication complex, is an attractive target for therapy. We
Chi-Pang Chuck et al.
PloS one, 5(10), e13197-e13197 (2010-10-16)
The 3C-like protease (3CL(pro)) of severe acute respiratory syndrome-coronavirus is required for autoprocessing of the polyprotein, and is a potential target for treating coronaviral infection. To obtain a thorough understanding of substrate specificity of the protease, a substrate library of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service