Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07745

Sigma-Aldrich

Diamond nanoparticles suspension

~1 % (w/w) solids

Synonym(s):

Nanodiamond

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.25

form

nanoparticles

concentration

~1 % (w/w) solids

density

1.005±0.005 g/mL at 20 °C

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandre Barras et al.
Nanoscale, 5(6), 2307-2316 (2013-02-12)
Bacterial attachment and subsequent biofilm formation on biotic surfaces or medical devices is an increasing source of infections in clinical settings. A large proportion of these biofilm-related infections are caused by Escherichia coli, a major nosocomial pathogen, in which the
Magnetic field imaging and more.
Daniel Evanko
Nature methods, 10(6), 468-468 (2013-07-20)
Diamond planets.
John Matson
Scientific American, 308(1), 12-12 (2013-01-25)
D Le Sage et al.
Nature, 496(7446), 486-489 (2013-04-27)
Magnetic imaging is a powerful tool for probing biological and physical systems. However, existing techniques either have poor spatial resolution compared to optical microscopy and are hence not generally applicable to imaging of sub-cellular structure (for example, magnetic resonance imaging)
Imaging: Magnetic bacteria on a diamond plate.
Mihály Pósfai et al.
Nature, 496(7446), 442-443 (2013-04-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service