Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07564

Sigma-Aldrich

P-Amidotriphosphoric acid mixed ammonium sodium salt solution

~ 1:3 molar ratio NH4+:Na+, ~0.5 M in H2O

Synonym(s):

AmTP, P-Amidotriphosphate mixed ammonium sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H6NO9P3 · xNH4+ · yNa+
Molecular Weight:
256.97 (free acid basis)
UNSPSC Code:
12352202
NACRES:
NA.25
Product 07564 is not currently sold in your country. Contact Technical Service

quality

~ 1:3 molar ratio NH4+:Na+

concentration

~0.5 M in H2O

storage temp.

2-8°C

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective alpha-Phosphorylation of Aldoses in Aqueous Solution Chemistry of alpha-aminonitriles, Part 29. Part 28: Ref. 1, Part 27: Ref. 2. This work was supported by the Skaggs Foundation. S.G. thanks the NASA NSCORT Exobiology program (La Jolla) for a postdoctoral fellowship.
Krishnamurthy et al.
Angewandte Chemie (International ed. in English), 39(13), 2281-2285 (2000-08-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service