Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07494

Sigma-Aldrich

L-Idaro-1,4-lactone

≥98.0% (TLC)

Synonym(s):

L-Idaric acid 1,4-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O7
CAS Number:
Molecular Weight:
192.12
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

assay

≥98.0% (TLC)

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O)C(=O)O[C@@H]1[C@@H](O)C(O)=O

InChI

1S/C6H8O7/c7-1-2(8)6(12)13-4(1)3(9)5(10)11/h1-4,7-9H,(H,10,11)/t1-,2-,3-,4+/m1/s1

InChI key

XECPAIJNBXCOBO-AJSXGEPRSA-N

Biochem/physiol Actions

L-Idaro-1,4-lactone is an inhibitor of α-L-idosiduronase, an enzyme that hydrolyzes iduronosidic linkages in the desulfated glycosaminoglycan dermatan.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service