Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06919

Sigma-Aldrich

Sulforhodamine 101 cadaverine trifluoroacetate salt

fluorescent probe for proteins

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H42N4O6S2 · C2F3O2
CAS Number:
Molecular Weight:
803.89
UNSPSC Code:
12352202
NACRES:
NA.32

fluorescence

λex 591 nm; λem 610 nm±10 nm in DMSO

storage temp.

2-8°C

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service