Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06840

Sigma-Aldrich

1-Methyltryptamine

≥95.0% (GC)

Synonym(s):

1-Methyl-1H-indole-3-ethanamine, 1-Methyl-3-(2-aminoethyl)indole, 2-(1-Methyl-1H-indol-3-yl)ethylamine, N1-Methyltryptamine, NMT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2
CAS Number:
Molecular Weight:
174.24
Beilstein/REAXYS Number:
6874
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Quality Level

assay

≥95.0% (GC)

form

liquid

color

light yellow

SMILES string

Cn1cc(CCN)c2ccccc12

InChI

1S/C11H14N2/c1-13-8-9(6-7-12)10-4-2-3-5-11(10)13/h2-5,8H,6-7,12H2,1H3

InChI key

CAAGZPJPCKMFBD-UHFFFAOYSA-N

Biochem/physiol Actions

Metabolite in N-methyltransferase metabolism of tryptamine and decarboxylation of 1-methyltryptophan.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Adkins et al.
Molecular pharmacology, 59(3), 514-523 (2001-02-17)
The serotonin (5-hydroxytryptamine, 5-HT) transporter (SERT) is responsible for the inactivation of synaptic 5-HT and is also a target for multiple psychostimulants. Despite the critical role of SERT in 5-HT inactivation and psychostimulant response, many aspects of the transporter's recognition
N-methylation of 1-methyltryptamines by indolethylamine N-methyltransferase.
R Lin et al.
Biochemical pharmacology, 24(11-12), 1239-1240 (1975-06-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service