Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06729

Sigma-Aldrich

Atto 488-dextran

mol wt 4 kDa, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

mol wt

4 kDa
Mw 3000-5000

fluorescence

λex 502 nm; λem 524 nm±10 nm in water

suitability

suitable for fluorescence

General description

A fluorescent derivative of a dextran fraction with an average molecular weight of approximately 4000-150′000.
Supplied as an orange powder which is readily soluble in water.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The changing point-spread function. Single-molecule-based super-resolution imaging.
Horrocks, M. H.; Palayret, M.; Klenerman, D.; Lee, S. F.
Histochemistry and Cell Biology, 141(6), 577-585 (2014)
Fast molecular tracking maps nanoscale dynamics of plasma membrane lipids.
Sahl, S. J.; et al.
Proceedings of the National Academy of Sciences of the USA, 107(15), 6829-6834 (2010)
Flexible Microdomain Specific Staining of Block Copolymers for 3D Optical Nanoscopy.
Ullal, Chaitanya K.; et al.
Macromolecules, 44(19), 7508-7510 (2011)
Saturated excitation microscopy for sub-diffraction-limited imaging of cell clusters.
Yamanaka, M.; et al.
Journal of Biomedical Optics, 18(12), 126002-126002 (2013)
2,2?-Thiodiethanol. A new water soluble mounting medium for high resolution optical microscopy.
Staudt, T.; et al.
Microscopy Research and Technique, 70(1), 1-9 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service