Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06479

Sigma-Aldrich

2-Anthracenesulfonyl chloride

BioReagent, suitable for fluorescence, ≥90.0% (HPLC)

Synonym(s):

2-Anthracene sulfonyl chloride, 2-Anthracenesulfonic acid chloride, Anthracene-2-sulfonic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9ClO2S
CAS Number:
Molecular Weight:
276.74
Beilstein/REAXYS Number:
2377545
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

assay

≥90.0% (HPLC)

form

powder

solubility

acetonitrile: soluble

fluorescence

λex 382 nm; λem 421 nm in acetonitrile (after derivatization with hexylamine)

suitability

suitable for fluorescence

SMILES string

ClS(=O)(=O)c1ccc2cc3ccccc3cc2c1

InChI

1S/C14H9ClO2S/c15-18(16,17)14-6-5-12-7-10-3-1-2-4-11(10)8-13(12)9-14/h1-9H

InChI key

FZAQROFXYZPAKI-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

In addition to the emission maximum at 421 nm, there are lower maxima at 405, 435 and 450 nm.

Other Notes

Derivatizing reagent with strong fluorescence[1]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J V Small et al.
Journal of cell science, 89 ( Pt 1), 21-24 (1988-01-01)
7-Diethylamino-3-(4-isothiocyanotophenyl)-4-methylcoumarin (CPITC) was coupled to amino-methyldithiolanophalloidin to produce a new phalloidin derivative, coumarin-phalloidin, fluorescent in the blue region of the spectrum. Coumarin-phalloidin binds to actin with around 100-fold less affinity than unconjugated phalloidin, but with enough avidity to make it

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service