Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05404

Sigma-Aldrich

AZCL-Amylose

substrate for α-amylase

Synonym(s):

Azurine-cross-linked amylose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

powder

storage temp.

2-8°C

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of cereal α-amylase: a new assay procedure.
McCleary, B. V. and Sheehan, H.
Journal of Cereal Science, 6, 237-251 (1987)
Determination of cereal alpha amylase using a commericially available dye-labeled substrate.
Barnes, W.C. and Blakeney, A.B.
Starch, 26, 193-197 (1974)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service