Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04675

Sigma-Aldrich

Peroxidase, Lyngbya sp., recombinant

≥0.6 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

specific activity

≥0.6 U/mg

storage temp.

−20°C

InChI

1S/H2O3/c1-3-2/h1-2H

InChI key

JSPLKZUTYZBBKA-UHFFFAOYSA-N

Unit Definition

1 Unit corresponds to the amount which oxidizes 1 μmol of ABTS per minute at 25°C and pH 5.0

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service