Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04533

Sigma-Aldrich

Esterase Pelobacter propionicus, recombinant

recombinant, expressed in E. coli, ≥0.5 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
UNSPSC Code:
12352204

recombinant

expressed in E. coli

form

crystalline
crystals
powder or flakes

specific activity

≥0.5 U/mg

storage temp.

−20°C

Application

Esterase Pelobacter propionicus, may be useful in the characterization of novel esterases. It has been used to aid the characterization of esterases from arctic sediment [1].

Biochem/physiol Actions

The esterase catalyzes the transesterification of 1-phenylethanol.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

1 U corresponds to the amount of enzyme which converts 1 μmol 4-nitrophenyl-L-acetate per minute at pH 7.5 and 30°C.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeong Ho Jeon et al.
Marine biotechnology (New York, N.Y.), 11(3), 307-316 (2008-09-25)
Two esterase genes (designated as estAT1 and estAT11, respectively) were cloned by activity-based screening of a fosmid library constructed with seashore sediment sample of the Arctic. The sequence analysis of the genes revealed that these esterase genes encoded proteins of
Marie C Fortin et al.
Drug metabolism and disposition: the biological fate of chemicals, 41(2), 326-331 (2012-12-12)
Studies on therapeutic drug disposition in humans have shown significant alterations as the result of pregnancy. However, it is not known whether pesticide metabolic capacity changes throughout pregnancy, which could affect exposure of the developing brain. We sought to determine
B Sànchez-Nogué et al.
Environmental science and pollution research international, 20(5), 3480-3488 (2012-12-06)
The common sole, Solea solea (Linneus, 1758), and the Senegalese sole, Solea senegalensis (Kaup, 1858), are two important commercial species that coexist in the NW Mediterranean. In order to assess the species' ability to respond to chemical insults, a comparison
Joshua P Lewis et al.
Pharmacogenetics and genomics, 23(1), 1-8 (2012-11-01)
Carboxylesterase 1 (CES1) is the primary enzyme responsible for converting clopidogrel into biologically inactive carboxylic acid metabolites. We genotyped a functional variant in CES1, G143E, in participants of the Pharmacogenomics of Anti-Platelet Intervention (PAPI) study (n=566) and in 350 patients
Sascha Obrowsky et al.
Journal of lipid research, 54(2), 425-435 (2012-12-12)
Adipose triglyceride lipase (ATGL) is the rate-limiting enzyme mediating triglyceride (TG) hydrolysis. The lack of ATGL results in TG accumulation in multiple tissues, underscoring the critical role of ATGL in maintaining lipid homeostasis. Recent evidence suggests that ATGL affects TG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service