Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03984

Sigma-Aldrich

Canada balsam solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

color

colorless to light yellow

suitability

suitable for microscopy

Looking for similar products? Visit Product Comparison Guide

Application

embedding medium for microscopy

Physical form

dissolved in xylene

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

78.8 °F

flash_point_c

26 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service