Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03899

Sigma-Aldrich

Prussian blue soluble

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Fe2KN6 · xH2O
CAS Number:
Molecular Weight:
306.89 (anhydrous basis)
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

solubility

water: 0.1 g/10 mL, blue to very deep blue

εmax

≥120 at 253-263 nm in water

suitability

suitable for microscopy

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[K+].[Fe++].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N
[K+].[Fe++].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.2Fe.K/c6*1-2;;;/q;;;;;;-3;+2;+1

InChI key

ZJSIHHBKYVAMOE-UHFFFAOYSA-N

Application

Prussian blue is not a dye but an inorganic pigment containing ferric ferrocyanide. Procedures have been published[1] for the demonstration of non-hemoglobin, cytoplasmic iron, according to Sundberg and Broman,[2] and for the demonstration of hemosiderin according to Highman[3] and from Lille and Fullmer[4] based the work originally done by Perle in 1867.[5] Prussian Blue is formed in situ in both of these methods. In combination with iron particles such as superparamagnetic iron oxide nanoparticles, Prussian blue may be used for tracking of transplanted cells such as mesenchymal stem cells.[6][7] Prussian blue may also be used to differentiate slow versus fast cycling cells by methods that monitor iron oxide particle labeling dilution thru cell division.[8]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang-Moon Lee et al.
Bioconjugate chemistry, 22(2), 186-192 (2011-01-20)
Oleic acid-conjugated chitosan (oleyl-chitosan) is a powerful platform for encapsulating oleic acid-decorated iron oxide nanoparticles (ION), resulting in a good magnetic resonance imaging (MRI) probe. Oleyl-chitosan could self-assemble into core-shell structures in aqueous solution and provide the effective core compartment
Dongbo Liu et al.
Acta biochimica et biophysica Sinica, 43(4), 301-306 (2011-02-25)
An accurate definition of clinical target volume (CTV) is essential for the application of radiotherapy in nasopharyngeal carcinoma (NPC) treatment. A novel epidermal growth factor receptor (EGFR)-targeting contrast agent (C225-USPIO) was designed by conjugating ultrasmall superparamagnetic iron oxide (USPIO) nanoparticles
A new modification of Perls's reaction for hemosiderin in tissues.
Highman, B.
Archives of Pathology, 33, 937-938 (1942)
In vivo and ex vivo MR imaging of slowly cycling melanoma cells.
Magnitsky S, Roesch A, Herlyn M, Glickson JD.
Magn. Reson. in Med., 66, 1362-1373 (2011)
Gerlinde Schmidtke-Schrezenmeier et al.
Cytotherapy, 13(8), 962-975 (2011-04-16)
Mesenchymal stromal cells (MSC) are the focus of research in regenerative medicine aiming at the regulatory approval of these cells for specific indications. To cope with the regulatory requirements for somatic cell therapy, novel approaches that do not interfere with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service