Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03065

Sigma-Aldrich

Lectin from Phytolacca americana-Atto 647N conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

description

carbohydrate specificity (GlcNAc)3

form

powder

fluorescence

λex 644 nm; λem 669 nm in PBS

General description

Lectins are ubiquitous proteins or glycoproteins with a high and specific affinity to sugar residues. They allow a sensitive detection of cellular carbohydrates, revealing subtle alteration in glycosylation between otherwise indistinguishable cells.
Atto-dyes have very bright fluorescent signals and high photostability, which enable a direct one-step tissue-binding protocol. Time-consuming multistage amplification procedures are not required for Atto-dye lectin conjugates.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanoscale organization of nicotinic aceylcholine receptors by stimulated emission depletion microscopy.
Kellner, R.R., et al.
Neuroscience, 144(1), 135-143 (2007)
A novel nanoscopic tool by combining AFM with STED microscopy.
Harke, B.; et al
Optical Nanoscopy, 1(1), 3-3 (2012)
Self-Calibrated Line-Scan STED-FCS to Quantify Lipid Dynamics in Model and Cell Membranes.
Benda, A.; Ma, Y.; Gaus, K.
Biophysical Journal, 108(3), 596-609 (2015)
STED nanoscopy combined with optical tweezers reveals protein dynamics on densely covered DNA.
Heller, I.; et al.
Nature Methods, 10(9), 910-916 (2013)
Naked Dense Bodies Provoke Depression.
Hallermann, S.; et al.
The Journal of Neuroscience, 30(43), 14340-14345 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service