Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01872

Supelco

CYPMPO

for ESR-spectroscopy, ≥97%

Synonym(s):

2-(5,5-Dimethyl-2-oxo-2λ5-[1,3,2]dioxaphosphinan-2-yl)-2-methyl-3,4-dihydro-2H-pyrrole 1-oxide, 5-(2,2-Dimethyl-1,3-propoxy cyclophosphoryl)-5-methyl-1-pyrroline N-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18NO4P
CAS Number:
Molecular Weight:
247.23
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

assay

≥97% (HPLC)
≥97%

storage temp.

−20°C

SMILES string

CC1(C)COP(=O)(OC1)C2(C)CCC=[N+]2[O-]

InChI

1S/C10H18NO4P/c1-9(2)7-14-16(13,15-8-9)10(3)5-4-6-11(10)12/h6H,4-5,7-8H2,1-3H3

InChI key

OSKIWEPJAIOTFB-UHFFFAOYSA-N

Application

CYPMPO is designed for radical detection. It is a free radical spin trap with excellent trapping capabilites toward hydroxyl and superoxide radicals in biological and chemical systems.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keiko Kobayashi et al.
Antioxidants (Basel, Switzerland), 9(3) (2020-03-20)
We investigated the effect of bisphenol A (BPA) on oxidative stress and tau-related proteins in adult rat brains. BPA (10 mg/L) was administered to rats for eight weeks through their drinking water. The reactive oxygen species (ROS) scavenging capacity for
Nobuyuki Endo et al.
Journal of clinical biochemistry and nutrition, 45(2), 193-201 (2009-10-02)
We have developed a simple ESR spin trapping based method for hydroxyl (OH) radical scavenging-capacity determination, using iron-free OH radical source. Instead of the widely used Fenton reaction, a short (typically 5 seconds) in situ UV-photolysis of a dilute hydrogen
Aki Hirayama et al.
Journal of clinical biochemistry and nutrition, 58(3), 167-173 (2016-06-04)
Kangen-karyu, a prescription containing six herbs, has been shown to achieve its pharmacological effect through oxidative stress-dependent pathways in animal models. The aim of this study is to investigate the relationship between the antioxidative effect and pharmacological mechanisms of Kangen-karyu

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service