Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01739

Sigma-Aldrich

Acrylamido buffer

≥98.0% (HPLC), pKa 10.3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

assay

≥98.0% (HPLC)

form

liquid

shelf life

limited shelf life

contains

~0.1% hydroquinone as stabilizer

refractive index

n20/D 1.479

pKa 

10.3

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Immobilized pH 2.5-11 gradients for 2-D electrophoresis[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Sinha et al.
Electrophoresis, 13(4), 210-214 (1992-04-01)
Extremely wide immobilized pH gradients, pH 2.5-11, for isoelectric separation of complex protein mixtures are described. These pH gradients are theoretically and practically the maximum that can be achieved at present with the available acrylamido buffers and titrants. Conditions are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service