Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00897

Sigma-Aldrich

2-[(1E)-2-(4-Azidophenyl)ethenyl]-6-nitrobenzothiazole

≥95.0% (HPLC)

Synonym(s):

(E)-6-Nitro-2-(4-azidostyryl)-1,3-benzothiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9N5O2S
CAS Number:
Molecular Weight:
323.33
Beilstein/REAXYS Number:
23580410
UNSPSC Code:
12352200
PubChem Substance ID:
Product 00897 is not currently sold in your country. Contact Technical Service

assay

≥95.0% (HPLC)

fluorescence

λex 440 nm; λem 570 nm±10 nm in DMF

storage temp.

−20°C

SMILES string

O=[N+]([O-])C1=CC=C2C(SC(/C=C/C3=CC=C(N=[N+]=[N-])C=C3)=N2)=C1

InChI

1S/C15H9N5O2S/c16-19-18-11-4-1-10(2-5-11)3-8-15-17-13-7-6-12(20(21)22)9-14(13)23-15/h1-9H/b8-3+

InChI key

IXXGAQKPFTVNHD-FPYGCLRLSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio F Melo Junior et al.
Life sciences, 257, 118138-118138 (2020-07-28)
Hypertension is a relevant sex and sex hormones-dependent risk factor where the cardiovascular and renal health of the population are concerned. Men experience greater losses of renal function (RF) than women, but the mechanisms remain somewhat unclear. Our goal was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service