Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88450

Supelco

Thioacetamide

ACS reagent, ≥99.0%

Synonym(s):

Ethanethioamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CSNH2
CAS Number:
Molecular Weight:
75.13
Beilstein/REAXYS Number:
506006
EC Number:
MDL number:
UNSPSC Code:
23151816
PubChem Substance ID:
Product 88450 is not currently sold in your country. Contact Technical Service

grade

ACS reagent

assay

≥99.0% (RT)
≥99.0%

form

crystals

ign. residue

≤0.05% (as SO4)

mp

108-112 °C (lit.)
111-114 °C

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

CC(N)=S

InChI

1S/C2H5NS/c1-2(3)4/h1H3,(H2,3,4)

InChI key

YUKQRDCYNOVPGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Used e.g. for the decomposition of amino acid copper complexes[1]

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Decomposition of amino acid copper complexes using thioacetamide.
U F Taylor et al.
International journal of peptide and protein research, 19(2), 158-161 (1982-02-01)
Chiung-Kuei Huang et al.
Hepatology (Baltimore, Md.), 57(4), 1550-1563 (2012-11-15)
Transplantation of bone marrow mesenchymal stem cells (BM-MSCs) has been considered as an alternative therapy, replacing liver transplantation in clinical trials, to treat liver cirrhosis, an irreversible disease that may eventually lead to liver cancer development. However, low survival rate
Suzy M Salama et al.
BMC complementary and alternative medicine, 13, 56-56 (2013-03-19)
Hepatology research has focused on developing traditional therapies as pharmacological medicines to treat liver cirrhosis. Thus, this study evaluated mechanisms of the hepatoprotective activity of Curcuma longa rhizome ethanolic extract (CLRE) on thioacetamide-induced liver cirrhosis in rats. The hepatoprotective effect
Peng Zhan et al.
Archives of pharmacal research, 35(6), 975-986 (2012-08-09)
In continuation of our endeavor to develop new, potent, selective and less toxic antiviral agents, a novel series of 2-(2-amino/chloro-4-(2,4-dibromophenyl) thiazol-5-ylthio)acetamide derivatives was synthesized via an expeditious route and evaluated for their anti-HIV activities against wild-type virus and clinically relevant
Mladen I Yovchev et al.
Hepatology (Baltimore, Md.), 59(1), 284-295 (2013-07-11)
Considerable progress has been made in developing antifibrotic agents and other strategies to treat liver fibrosis; however, significant long-term restoration of functional liver mass has not yet been achieved. Therefore, we investigated whether transplanted hepatic stem/progenitor cells can effectively repopulate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service