Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

80640

Sigma-Aldrich

Piperidine

puriss. p.a., ≥99.0% (GC/T)

Synonym(s):

Hexahydropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15
Beilstein/REAXYS Number:
102438
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21

vapor density

3 (vs air)

vapor pressure

23 mmHg ( 20 °C)

grade

puriss. p.a.

assay

≥99.0% (GC/T)

form

liquid

impurities

≤0.3% pyridine (GC)
≤0.3% water

evapn. residue

≤0.02%

refractive index

n20/D 1.452 (lit.)
n20/D 1.453

bp

106 °C (lit.)

mp

−13 °C (lit.)

solubility

organic solvents: soluble(lit.)
water: miscible(lit.)

density

0.862 g/mL at 20 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

C1CCNCC1

InChI

1S/C5H11N/c1-2-4-6-5-3-1/h6H,1-5H2

InChI key

NQRYJNQNLNOLGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Piperidine may be employed as protective and structure-directing agent in the post synthesis of MCM (Mobil Composition of Matter)-49 zeolites.[1]

Other Notes

Sales restrictions may apply

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

60.8 °F - closed cup

flash_point_c

16 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rubiralta M, et al.
Piperidine: Structure, Preparation, Reactivity, and Synthetic Applications of Piperidine and its Derivatives, 43, 2-2 (2013)
Moessbauer spectra of some porphyrin complexes with pyridine, piperidine, and imidazole.
Epstein LM, et al.
Inorganic Chemistry, 6(9), 1720-1724 (1967)
Simultaneous recrystallization and post-synthesis of MCM-49 zeolite by TEAOH/piperidine: Implication on the acidity and catalytic alkylation performance.
Shi Y, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 220, 7-15 (2016)
Rubiralta M, et al.
Piperidine: Structure, Preparation, Reactivity, and Synthetic Applications of Piperidine and its Derivatives, 43, 14-14 (2013)
Rubiralta M, et al.
Piperidine: Structure, Preparation, Reactivity, and Synthetic Applications of Piperidine and its Derivatives, 43, 10-10 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service