Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77370

Sigma-Aldrich

Petroleum

special, ~18% aromatics basis, bp 180-220 °C

Synonym(s):

White Spirit high-boiling

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352401
bp:
180-220 °C

form

liquid

contains

~18% aromatics

concentration

~18% (aromatics)

refractive index

n20/D 1.441

bp

180-220 °C

density

0.79 g/mL at 20 °C

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Muta. 1B - STOT RE 1

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

141.8 °F - closed cup

flash_point_c

61 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 267-267 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 660-660 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 916-916 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service