Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

676799

Sigma-Aldrich

Hexane, mixture of isomers

≥98.5%, ACS reagent

Synonym(s):

Hexanes, Isohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
Beilstein/REAXYS Number:
1730733
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
ACS reagent
assay:
≥98.5%
bp:
68-70 °C (lit.)

Product Name

Hexane, mixture of isomers, ACS reagent, ≥98.5%

grade

ACS reagent

vapor density

3 (vs air)

assay

≥98.5%

autoignition temp.

437 °F

expl. lim.

7.5 %

dilution

(for analytical testing)

impurities

Mixture of hexane isomers plus methylcyclopentane
Thiophene, passes test
≤0.0003 meq/g water-soluble titr. acid
≤0.005% S compounds

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.379 (lit.)

bp

68-70 °C (lit.)

mp

−95 °C (lit.)

density

0.672 g/mL at 25 °C (lit.)
0.672 g/mL at 25 °C

InChI

1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3

InChI key

AFABGHUZZDYHJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-9.4 °F

flash_point_c

-23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service