Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65141

Sigma-Aldrich

Nonafluorobutyl methyl ether - trans-1,2-Dichloroethylene mixture

puriss., 1:1 azeotrope, fluorochlorinated hydrocarbon substitute solvent, ≥99.0% (total assay of azeotrope, GC)

Synonym(s):

HFE-71 DE, Methoxynonafluorobutane - trans-1,2-Dichloroethylene 1:1 azeotrope, Methoxyperfluorobutane - trans-1,2-Dichloroethylene 1:1 azeotrope, Nonafluoromethoxybutane - trans-1,2-Dichloroethylene 1:1 Azeotrope

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3F9O · C2H2Cl2
CAS Number:
Molecular Weight:
347.01
UNSPSC Code:
12352101
PubChem Substance ID:
assay:
≥99.0% (total assay of azeotrope, GC)
technique(s):
GC/GC: suitable
bp:
41 °C (lit.)

grade

puriss.

assay

≥99.0% (total assay of azeotrope, GC)

form

liquid

technique(s)

GC/GC: suitable

refractive index

n20/D 1.354

bp

41 °C (lit.)

SMILES string

Cl\C=C\Cl.COC(F)(F)C(F)(F)C(F)(F)C(F)(F)F.COC(F)(F)C(F)(C(F)(F)F)C(F)(F)F

InChI

1S/2C5H3F9O.C2H2Cl2/c1-15-5(13,14)3(8,9)2(6,7)4(10,11)12;1-15-5(13,14)2(6,3(7,8)9)4(10,11)12;3-1-2-4/h2*1H3;1-2H/b;;2-1+

InChI key

HHJKFLXMNOPRIM-WXXKFALUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solvent which has no ozone-degrading activity and is only slightly toxic

hcodes

pcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service