60243
Potassium formate
purum p.a., ≥99.0% (NT)
Synonym(s):
Formic acid potassium salt
About This Item
grade
purum p.a.
assay
≥99.0% (NT)
form
powder, crystals or chunks
mp
165-168 °C (lit.)
anion traces
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg
cation traces
Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
Na: ≤5000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg
InChI
1S/CH2O2.K/c2-1-3;/h1H,(H,2,3);/q;+1/p-1
InChI key
WFIZEGIEIOHZCP-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service