Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

562602

Sigma-Aldrich

Cesium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form:
solid

form

solid

Quality Level

mp

645 °C (lit.)

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

application

Used for the preparation of electrically conducting glasses.Used to make solutions for the separation of RNA from DNA by density gradient centrifugation.

Legal Information

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuan A Ho et al.
Langmuir : the ACS journal of surfaces and colloids, 28(2), 1256-1266 (2011-12-14)
All-atom molecular dynamics simulations were conducted to study the dynamics of aqueous electrolyte solutions confined in slit-shaped silica nanopores of various degrees of protonation. Five degrees of protonation were prepared by randomly removing surface hydrogen atoms from fully protonated crystalline
Shengyuan Ding et al.
Journal of neurophysiology, 106(6), 3019-3034 (2011-09-02)
GABA projection neurons (GABA neurons) in the substantia nigra pars reticulata (SNr) and dopamine projection neurons (DA neurons) in substantia nigra pars compacta (SNc) have strikingly different firing properties. SNc DA neurons fire low-frequency, long-duration spikes, whereas SNr GABA neurons
Raphaele Germi et al.
The Journal of general virology, 93(Pt 8), 1769-1773 (2012-05-18)
Epstein-Barr virus (EBV), a gammaherpesvirus, infects >90 % of the world's population. Primary infection by EBV can lead to infectious mononucleosis, and EBV persistence is associated with several malignancies. Despite its importance for human health, little structural information is available on
Takafumi Yamamoto et al.
Inorganic chemistry, 50(22), 11787-11794 (2011-10-25)
The rock salt (B1) structure of binary oxides or chalcogenides transforms to the CsCl (B2) structure under high pressure, with critical pressures P(s) depending on the cation to anion size ratio (R(c)/R(a)). We investigated structural changes of A(2)MO(3) (A =
Therese Abrahamsson et al.
Neuron, 73(6), 1159-1172 (2012-03-27)
Interneurons are critical for neuronal circuit function, but how their dendritic morphologies and membrane properties influence information flow within neuronal circuits is largely unknown. We studied the spatiotemporal profile of synaptic integration and short-term plasticity in dendrites of mature cerebellar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service