Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51127

Sigma-Aldrich

Activated Charcoal Norit®

Norit® RB4C, steam activated, rod

Synonym(s):

Charcoal activated Norit®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Beilstein/REAXYS Number:
4360473
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:

form

rod

quality

steam activated

manufacturer/tradename

Norit® RB4C

description

18-23% (butane adsorption (p/po = 0.1))

diam.

~4 mm

loss

≤5% loss on drying

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Norit is a registered trademark of Norit N.V.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of activated charcoal for the removal of patulin from cider.
Sands DC, et al.
Applied and Environmental Microbiology, 32(3), 388-391 (1976)
Diazepam stability in wastewater and removal by advanced membranes technology, activated carbon and micelle-clay complex
Bufo SA, et al.
Desalination and Water Treatment (2015)
Lithium recovery from aqueous solution by sorption/desorption
Lemaire J, et al.
Hydrometallurgy, 143, 1-11 (2014)
About reactions occurring during chemical activation with hydroxides.
Lillo-Rodenas MA, et al.
Carbon, 42(7), 1371-1375 (2004)
Activated carbon for gas separation and storage
Sircar S, et al.
Carbon, 34(1), 1-12 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service