Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

402818

Sigma-Aldrich

Ammonium aluminum sulfate dodecahydrate

ACS reagent, ≥98%

Synonym(s):

Aluminum ammonium sulfate dodecahydrate, Ammonium alum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlNH4(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
453.33
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

assay

≥98%
98.0-102.0% (ACS specification)

impurities

≤0.005% insolubles

mp

93.5 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%

cation traces

Ca: ≤0.05%
Fe: ≤0.001%
K: ≤0.05%
Na: ≤0.01%
heavy metals (as Pb): ≤0.001%

SMILES string

[Al+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H][N+]([H])([H])[H].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Al.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3

InChI key

WZUKKIPWIPZMAS-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium aluminum sulfate dodecahydrate commonly called as ammonium alum is widely used in water purification, cosmetic formulation, fertilizers and food preservation. Its thermal decomposition has been studied.[1] It has high latent heat of 251 kJ kg-1 with a potential use in the heating system.[2] Its utility as a flame-retardant for cotton fabric has been analyzed.[3]

Application

Ammonium aluminum sulfate dodecahydratemay be used in the preparation of Alum (Al(OH)3) gel.[4]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preventive and curative glycoside kaempferol treatments attenuate the TH2-driven allergic airway disease.
Medeiros KCP, et al.
International Immunopharmacology, 9(13), 1540-1548 (2009)
Thermogravimetry of deposited ammonium aluminum sulfate dodecahydrate used as flame-retardant for cotton fabrics.
Mostashari SM and Mostashari SZ.
Cellulose Chemistry and Technology, 43(9), 455-455 (2009)
Thermal characterization of ammonium alum.
Lopez-Beceiro J, et al.
Journal of Thermal Analysis and Calorimetry, 104(1), 127-130 (2010)
Flow and heat transfer characteristics of ammonium alum hydrate slurries.
Suzuki H, et al.
International Journal of Refrigeration, 36(1), 81-87 (2013)
Mutsuko Hirata-Koizumi et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(9), 1948-1959 (2011-05-17)
Aluminium ammonium sulfate (AAS) was tested for reproductive/developmental toxicity in a two-generation study. Male and female rats were continuously given AAS in drinking water at 0, 50, 500 or 5000 ppm. Water consumption was decreased in all AAS-treated groups, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service