Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

34872

Sigma-Aldrich

1,2-Dichloroethane

suitable for HPLC, ≥99.8%

Synonym(s):

Ethylene chloride, Ethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Beilstein/REAXYS Number:
605264
EC Number:
MDL number:
UNSPSC Code:
41116105
eCl@ss:
39050106
PubChem Substance ID:
NACRES:
NA.04

vapor density

3.4 (20 °C, vs air)

vapor pressure

87 mmHg ( 25 °C)

assay

≥99.8%

form

liquid

autoignition temp.

775 °F

expl. lim.

15.6 %

technique(s)

HPLC: suitable

impurities

≤0.0005% non-volatile matter
≤0.001% free acid (as HCl)
≤0.02% water (Karl Fischer)

transmittance

230 nm, ≥25%
245 nm, ≥90%
270 nm, ≥98%

refractive index

n20/D 1.444 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

density

1.256 g/mL at 25 °C (lit.)

application(s)

food and beverages

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dichloroethane (ethylene dichloride, DCE) is a solvent. It is one of the degradation by-product released from vinyl chloride manufacturing industries (EDC-tar). Its mutagenic activity has been tested on Salmonella typhimurium TA 1535.[1] Its catalytic degradation over titania/silica (acid catalysts) has been investigated.[2]

Application

1,2-Dichloroethane (ethylene dichloride, DCE) has been used for the chemical doping during the fabrication of Cl-doped few-layer WS2 or MoS2 back gate field-effect transistors (FETs).[3] It may be employed as an electron acceptor in the culture medium of Dehalococcoides ethenogenes 195.[4] It may be employed as model compound for chlorinated organics in various studies.[2]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sanae Sakami et al.
Methods in molecular biology (Clifton, N.J.), 1834, 311-332 (2018-10-17)
Immuno-EM is a method that can determine the localization of a protein in a tissue at the ultrastructural level. Generally, membrane structures in immuno-EM specimens have very low contrast because fixation is performed without osmium. Here, by using high-angle annular
Lingming Yang et al.
Nano letters, 14(11), 6275-6280 (2014-10-14)
Low-resistivity metal-semiconductor (M-S) contact is one of the urgent challenges in the research of 2D transition metal dichalcogenides (TMDs). Here, we report a chloride molecular doping technique which greatly reduces the contact resistance (Rc) in the few-layer WS2 and MoS2.
Decomposition of 1, 2-dichloroethane on titanium dioxide/silica.
Imamura S, et al.
Industrial & Engineering Chemistry Research, 28(10), 1449-1452 (1989)
U Rannug et al.
Chemico-biological interactions, 20(1), 1-16 (1978-01-01)
One of the main components in the waste products from vinyl chloride industries (EDC-tar), is ethylene dichloride (1,2-dichloroethane). This compound has been tested for mutagenicity on Salmonella typhimurium TA 1535. It is concluded that 1,2-dichloroethane gives a weak direct mutagenic
X Maymó-Gatell et al.
Applied and environmental microbiology, 65(7), 3108-3113 (1999-07-02)
"Dehalococcoides ethenogenes" 195 can reductively dechlorinate tetrachloroethene (PCE) completely to ethene (ETH). When PCE-grown strain 195 was transferred (2% [vol/vol] inoculum) into growth medium amended with trichloroethene (TCE), cis-dichloroethene (DCE), 1,1-DCE, or 1,2-dichloroethane (DCA) as an electron acceptor, these chlorinated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service