Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34870

Sigma-Aldrich

Ethyl alcohol, pure

≥99.8% (GC), suitable for HPLC, absolute, sales not in Germany

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein/REAXYS Number:
1718733
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.05
assay:
≥99.8% (GC)
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
78 °C (lit.)
vapor pressure:
44.6 mmHg ( 20 °C)

Product Name

Ethanol, absolute, suitable for HPLC, sales not in Germany1, ≥99.8% (GC)

vapor density

1.59 (vs air)

Quality Level

vapor pressure

44.6 mmHg ( 20 °C)

grade

absolute

assay

≥99.8% (GC)

form

liquid

autoignition temp.

683 °F

quality

sales not in Germany1

expl. lim.

19 %, 60 °F

technique(s)

HPLC: suitable

impurities

≤0.001% free acid (as CH3COOH)
≤0.001% non-volatile matter
≤0.2% water (Karl Fischer)
≤1 ppb fluorescence (quinine) at 254 nm
≤1 ppb fluorescence (quinine) at 365 nm

transmittance

210 nm, ≥20%
225 nm, ≥50%
240 nm, ≥80%
260 nm, ≥98%

refractive index

n20/D 1.3600 (lit.)

pH

7.0 (20 °C, 10 g/L)

bp

78 °C (lit.)

mp

−114 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethanol is a highly combustible colorless liquid. It is highly reactive in nature. It oxidizes to afford acetaldehyde or acetic acid. It undergoes reaction with carboxylic acid and derivatives to form esters, with aldehydes and ketones to afford acetals and ketals, respectively.[1] The monitoring of ethanol levels is very important in control of fermentation processes and in certification of alcoholic beverages.[2]

Application

Ethanol has been used in the mobile phase for the quantification of coenzyme Q10 (ubiquinone) by HPLC.[3] It has been used to compose the receptor medium to investigate the release profiles of the various pharmaceutical formulations of ketoconazole drug and their in vitro diffusion studies.[4]

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ketoconazole in topical pharmaceutical formulations. The influence of the receptor media on the in vitro diffusion kinetics.
Mitu MA, et al.
Farmacia, 59(3), 358-366 (2011)
Eagleson M.
Concise Encyclopedia Chemistry, 384-384 (1994)
Ufuk Cobanoglu et al.
Asian Pacific journal of cancer prevention : APJCP, 12(6), 1399-1403 (2011-12-01)
Early diagnosis and prevention is very important for lung cancer patients. Previous studies have emphasized that the level of coenzyme Q10 (CoQ10), present primarily in mitochondria, decreases with age and is low in patients with chronic diseases. Our goal was
A new oxidase-peroxidase kit for ethanol assays in alcoholic beverages.
Gonchar MV, et al.
Food Technology and Biotechnology, 39(1), 37-42 (2001)
Cem Kuscu et al.
Nature biotechnology, 32(7), 677-683 (2014-05-20)
RNA-guided genome editing with the CRISPR-Cas9 system has great potential for basic and clinical research, but the determinants of targeting specificity and the extent of off-target cleavage remain insufficiently understood. Using chromatin immunoprecipitation and high-throughput sequencing (ChIP-seq), we mapped genome-wide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service