Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

342777

Sigma-Aldrich

Magnesium oxide

fused, chips, -4 mesh, 99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:

assay

99.9% trace metals basis

form

chips
fused

reaction suitability

reagent type: catalyst
core: magnesium

particle size

-4 mesh

mp

2852 °C (lit.)

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for vacuum deposition.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication of β-TCP foam: Effects of magnesium oxide as phase stabilizer on its properties.
Nikaido T, et al.
Ceramics International, 41(10), 14245-14250 (2015)
Synthesis of ethylene and ethane by partial oxidation of methane over lithium-doped magnesium oxide.
Ito T and Lunsford JH.
Nature, 721-722 (1985)
Eagleson M.
Concise Encyclopedia Chemistry, 495-495 (1994)
Magnesium oxide-catalysed reaction of carbon dioxide with an epoxide with retention of stereochemistry.
Yano T, et al.
Chemical Communications (Cambridge, England), 12, 1129-1130 (1997)
Magdalena Maciejewska et al.
Journal of chromatography. A, 1257, 141-148 (2012-08-22)
Inverse gas chromatography (IGC) was applied at infinite dilution to evaluate the surface properties of calcium and magnesium oxide nanoparticles and the effect of surface grafted unsaturated carboxylic acid on the nanopowder donor-acceptor characteristics. The dispersive components (γ(s)(D)) of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service