Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

319309

Sigma-Aldrich

Potassium chloride solution

in H2O (saturated)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Beilstein/REAXYS Number:
1711999
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solution

concentration

in H2O (saturated)
≥25% (silver nitrate titration)

pH

5-8 (25 °C, 260 g/L)

density

1.175 g/mL at 25 °C

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hanne Sørup Tastesen et al.
Amino acids, 46(7), 1659-1671 (2014-03-25)
High-protein diets induce alterations in metabolism that may prevent diet-induced obesity. However, little is known as to whether different protein sources consumed at normal levels may affect diet-induced obesity and associated co-morbidities. We fed obesity-prone male C57BL/6J mice high-fat, high-sucrose
Sandra S Wise et al.
Toxicology and applied pharmacology, 279(2), 113-118 (2014-06-22)
Sea turtles are a charismatic and ancient ocean species and can serve as key indicators for ocean ecosystems, including coral reefs and sea grass beds as well as coastal beaches. Genotoxicity studies in the species are absent, limiting our understanding
Yuanyuan Qu et al.
Nucleic acids research, 41(10), 5263-5272 (2013-04-13)
Bacterial nucleoid-associated proteins, such as H-NS-like proteins in Enterobacteriaceae, are abundant DNA-binding proteins that function in chromosomal DNA organization and gene transcription regulation. The Mycobacterium tuberculosis Lsr2 protein has been proposed to be the first identified H-NS analogue in Gram-positive
David A Story et al.
Anesthesiology, 119(3), 569-575 (2013-04-20)
In an incidental finding, during a study of plasma chemistry after crystalloid infusion, participants reported subjective cognitive changes, particularly slower thinking, after saline but not Hartmann's (Ringer's lactate) solution. The authors tested the hypothesis that saline infusion would produce greater
Garrett A Kaas et al.
Neuron, 79(6), 1086-1093 (2013-09-21)
Dynamic changes in 5-methylcytosine (5mC) have been implicated in the regulation of gene expression critical for consolidation of memory. However, little is known about how these changes in 5mC are regulated in the adult brain. The enzyme methylcytosine dioxygenase TET1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service