Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31440

Sigma-Aldrich

Sodium nitrate

puriss. p.a., ACS reagent, reag. ISO, ≥99.5%

Synonym(s):

Chile salpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaNO3
CAS Number:
Molecular Weight:
84.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99.5%

grade

ACS reagent
puriss. p.a.

agency

reag. ISO

assay

≥99.5%

impurities

≤0.002% ammonium (NH4)
≤0.005% in water insoluble matters

pH

5.5-8.0 (20 °C, 5%)

mp

306 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
iodate (IO3-): ≤5 mg/kg
nitrite (NO2-): ≤10 mg/kg
phosphate (PO43-): ≤2 mg/kg
sulfate (SO42-): ≤30 mg/kg

cation traces

Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤2 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].[O-][N+]([O-])=O

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium nitrate is an inorganic sodium salt. Sodium nitrate (Chile saltpeter, NaNO3) is found naturally in abundant amounts in paleogeochemical ores in northern Chile. Sodium nitrate can be obtained from these ores by mining and leaching. NaNO3 can be used as a fertilizer in agricultural fields.[1]

Application

Sodium nitrate may be employed in polyol process for the modification of morphological features of Pt nanoparticles.[2] It may be employed for the modification of natural phosphate (NP) to afford natural phosphate doped with sodium nitrate (NaNO3/NP). NaNO3/NP was used as a catalyst during the synthesis of chalcones.[3]

pictograms

Flame over circleExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcined sodium nitrate/natural phosphate: an extremely active catalyst for the easy synthesis of chalcones in heterogeneous media.
Sebti S, et al.
Tetrahedron Letters, 42(45), 7953-7955 (2001)
E T Urbansky et al.
Environmental pollution (Barking, Essex : 1987), 112(3), 299-302 (2001-04-09)
Paleogeochemical deposits in northern Chile are a rich source of naturally occurring sodium nitrate (Chile saltpeter). These ores are mined to isolate NaNO3 (16-0-0) for use as fertilizer. Coincidentally, these very same deposits are a natural source of perchlorate anion
Polyol synthesis of platinum nanoparticles: control of morphology with sodium nitrate.
Herricks T, et al.
Nano Letters, 4(12), 2367-2371 (2004)
Suna Choi et al.
Journal of controlled release : official journal of the Controlled Release Society, 131(1), 70-76 (2008-08-12)
One of the most important requirements for non-viral gene delivery systems is the ability to mediate high levels of gene expression with low toxicity. After the DNA/vector complexes are taken up by cells through endocytosis, DNA is typically contained within
Nicole K Richards et al.
Environmental science & technology, 46(19), 10447-10454 (2012-04-18)
Nitrate and halide ions coexist in a number of environmental systems, including sea salt particles, the Arctic snowpack, and alkaline dry lakes. However, little is known about potential synergisms between halide and nitrate ions. The effect of sea salt on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service