Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31268

Sigma-Aldrich

Potassium disulfite

puriss. p.a., ≥96% (iodometric), powder

Synonym(s):

Potassium metabisulfite, Potassium pyrosulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2S2O5
CAS Number:
Molecular Weight:
222.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥96% (iodometric)
form:
powder

grade

puriss. p.a.

assay

≥96% (iodometric)

form

powder

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

As: ≤1 mg/kg
Cu: ≤10 mg/kg
Fe: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg

SMILES string

[K+].[K+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2K.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

RWPGFSMJFRPDDP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

supp_hazards

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Mathison et al.
Chest, 87(1 Suppl), 50S-54S (1985-01-01)
Several types of reactions to drugs and chemicals may precipitate or perpetuate asthmatic relapse. This review focuses on reactions to aspirin and sulfites. Approximately 40 percent of patients with rhinosinusitis, nasal polyps, and asthma and 5 to 10 percent of
S Raisuddin et al.
Food additives and contaminants, 8(6), 707-712 (1991-11-01)
The occurrence of aflatoxins in market betel nut samples was studied. It was observed that several betel nut samples were infested with aflatoxin-producing fungus, Aspergillus flavus. Out of 32 samples collected from various places, 12 were positive for aflatoxin. Aflatoxin
Carcinogenicity test of potassium metabisulfite in mice.
T Tanaka et al.
Ecotoxicology and environmental safety, 3(4), 451-453 (1979-12-01)
E Pastorkova et al.
International journal of food microbiology, 161(3), 209-213 (2013-01-22)
This paper investigates the in vitro antimicrobial potential of 15 grape phenolic compounds of various chemical classes (phenolic acids, stilbenes and flavonoids) using the broth microdilution method against yeasts and acetic acid bacteria frequently occurring in deteriorated wine. Pterostilbene (MICs=32-128
M Takahashi et al.
Japanese journal of cancer research : Gann, 77(2), 118-124 (1986-02-01)
Ethanol, potassium metabisulfite, formaldehyde and hydrogen peroxide were tested for tumor-promoting activity in a two-stage stomach carcinogenesis experiment. Male outbred Wistar rats were given N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) in the drinking water (100 mg/liter) and a diet supplemented with 10% sodium chloride

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service