Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31260

Supelco

Potassium bisulfate

puriss. p.a., ≥99%

Synonym(s):

Potassium hydrogen sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KHSO4
CAS Number:
Molecular Weight:
136.17
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99%
form:
solid

grade

puriss. p.a.

assay

≥99%

form

solid

impurities

≤0.001% total nitrogen (N)

mp

214 °C (lit.)

density

2.32 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
phosphate (PO43-): ≤5 mg/kg

cation traces

Al: ≤20 mg/kg
As: ≤0.5 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤5 mg/kg
Na: ≤50 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[K+].OS([O-])(=O)=O

InChI

1S/K.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+1;/p-1

InChI key

CHKVPAROMQMJNQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Li et al.
Chemosphere, 188, 139-147 (2017-09-08)
Electrochemistry coupled with granulated activated carbon catalysis of peroxymonosulfate (electro/GAC/PMS) as a novel wastewater treatment process was performed for the degradation of Acid Orange 7 (AO7) in aqueous solution. The decolorization of AO7 was compared under different permutations and combinations
Emily J Dennis et al.
Nature, 562(7725), 119-123 (2018-09-28)
DEET (N,N-diethyl-meta-toluamide) is a synthetic chemical identified by the US Department of Agriculture in 1946 in a screen for repellents to protect soldiers from mosquito-borne diseases1,2. Since its discovery, DEET has become the world's most widely used arthropod repellent and
Sunghee Lee et al.
Langmuir : the ACS journal of surfaces and colloids, 26(12), 9556-9564 (2010-02-18)
In this work, we have investigated the effect of potassium salts of different anions upon the crystal nucleation of K(2)SO(4) as interfacially templated by a surfactant monolayer of 1-octadecylamine (ODA), in an aqueous microdroplet system bounded by a liquid-liquid interface
Michael B Ellis et al.
Journal of insect physiology, 54(12), 1516-1521 (2008-09-30)
Terrestrial organisms need to limit evaporation from their bodies in order to maintain a homeostatic water balance. Owing to a large surface to volume ratio, arthropods are particularly susceptible to desiccation and have evolved behavioural and physiological mechanisms to conserve
Tian-Li Ma et al.
BMC plant biology, 12, 161-161 (2012-09-12)
Potassium (K+) is an important nutrient ion in plant cells and plays crucial roles in many plant physiological and developmental processes. In the natural environment, K+ deficiency is a common abiotic stress that inhibits plant growth and reduces crop productivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service