Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31255

Sigma-Aldrich

Potassium dichromate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%

Synonym(s):

Potassium bichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2Cr2O7
CAS Number:
Molecular Weight:
294.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99.8%
form:
solid

grade

ACS reagent
puriss. p.a.

agency

USP/NF
reag. ISO
reag. Ph. Eur.

assay

≥99.8%

form

solid

impurities

≤0.005% water insolubles

loss

≤0.05% loss on drying, 105 °C

mp

398 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤20 mg/kg
Cu: ≤10 mg/kg
Fe: ≤10 mg/kg
Na: ≤100 mg/kg
Pb: ≤50 mg/kg

SMILES string

[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2K.7O/q;;2*+1;;;;;;2*-1

InChI key

KMUONIBRACKNSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

target_organs

Cardio-vascular system, Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo Molina-Jijón et al.
Toxicology, 291(1-3), 93-101 (2011-11-26)
Deferoxamine (DFO) is a recognized iron chelator which has been shown to exert nephroprotection in models of toxic nephropathies. In the present work the potential protective effects of DFO against Cr(VI)-induced nephrotoxicity and oxidant stress were evaluated. Rats were injected
Bour-Jr Wang et al.
Journal of the Formosan Medical Association = Taiwan yi zhi, 110(12), 775-779 (2012-01-18)
Occupational dermatitis among cement workers is a major occupational health concern. The two most important occupational hazards for cement workers are irritant and allergic cement contact dermatitis. The objective of this study was to investigate the severity of occupational cement
Shafaqat Ali et al.
Environmental toxicology and chemistry, 32(10), 2234-2239 (2013-06-19)
A hydroponic experiment was carried out to examine the effect of hydrogen sulfide (H2 S) in alleviating chromium (Cr) stress in barley. A 2-factorial design with 6 replications was selected, including 3 levels of NaHS (0 μM, 100 μM, and
Lubna Shakir et al.
Environmental toxicology and pharmacology, 34(2), 180-191 (2012-04-24)
The problem of water pollution acquires greater relevance in the context of a developing agrarian economy like Pakistan. Even though, the leather industry is a leading economic sector in Pakistan, there is an increasing environmental concern regarding tanneries because they
Ashish K Mishra et al.
Environmental toxicology, 27(7), 415-422 (2012-06-19)
Exposure to sublethal hexavalent chromium (as 2 and 4 mg L⁻¹ potassium dichromate for 1 and 2 months) during late preparatory to mid prespawning phase of annual reproductive cycle severely affected the pituitary-ovarian axis of a teleost Channa punctatus. Gonadosomatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service