Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31127

Sigma-Aldrich

Barium hydroxide octahydrate

puriss. p.a., reag. ISO, reag. Ph. Eur., ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(OH)2 · 8H2O
CAS Number:
Molecular Weight:
315.46
EC Number:
MDL number:
UNSPSC Code:
12352305
PubChem Substance ID:
assay:
≥98%

agency

USP/NF
reag. ISO
reag. Ph. Eur.

grade

puriss. p.a.

assay

≥98%

impurities

≤0.005% insoluble in acid
≤2% BaCO3

mp

78 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfide (S2-): ≤5 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤20 mg/kg
Na: ≤50 mg/kg
Pb: ≤5 mg/kg
Sr: ≤1.5%
Zn: ≤5 mg/kg

SMILES string

O.O.O.O.O.O.O.O.O[Ba]O

InChI

1S/Ba.10H2O/h;10*1H2/q+2;;;;;;;;;;/p-2

InChI key

ZUDYPQRUOYEARG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyzes the β-elimination of phosphoserine residues.

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of barium hydroxide octahydrate Ba(OH)2-8H2O.
Manohar H and Ramaseshan S.
Zeitschrift fur Kristallographie, 119(1-6), 357-374 (1964)
Duk Kyu Lee et al.
Journal of nanoscience and nanotechnology, 10(2), 1361-1366 (2010-04-01)
BaTiO3 nano-particles with a size range of 20-70 nm were synthesized at 70 degrees C through a simple wet-chemical reactions between barium hydroxide octahydrate (Ba(OH)2 x 8H2O) and meta titanic acid (H2TiO3) in ethanol/water mixed solvent systems. It was found
Sjoerd A Veldhuis et al.
Inorganic chemistry, 53(24), 13188-13196 (2014-11-26)
Nanocrystalline barium titanate (8-10 nm crystallite size) was prepared at temperatures of 23-78 °C through reaction of a modified titanium alkoxide precursor in benzyl alcohol with barium hydroxide octahydrate. The room temperature formation of a perovskite phase from solution is
B J Verhaeghe et al.
Clinical chemistry, 34(6), 1077-1083 (1988-06-01)
This is a new method for isolating and concentrating urinary organic acids before gas chromatography--mass spectrometry. Sulfate and phosphate anions are removed by precipitation with Ba(OH)2, and the urine pH is adjusted to 8-8.5. The sample is then applied onto
Eagleson M.
Concise Encyclopedia Chemistry, 116-116 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service