31120
Ammonium thiocyanate
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (AT)
Synonym(s):
Ammonium rhodanide
About This Item
Recommended Products
grade
ACS reagent
puriss. p.a.
agency
reag. ISO
reag. Ph. Eur.
assay
≥99% (AT)
impurities
≤0.0005% heavy metals (as Pb)
≤0.005% I red. matter (as SO2)
ign. residue
≤0.02% (as SO4)
pH
4.5-6.0 (20 °C, 5%)
mp
152-154 °C (lit.)
anion traces
chloride (Cl-): ≤40 mg/kg
sulfate (SO42-): ≤50 mg/kg
sulfide (S2-): ≤10 mg/kg
cation traces
Cu: ≤5 mg/kg
Fe: ≤1 mg/kg
SMILES string
N.SC#N
InChI
1S/CHNS.H3N/c2-1-3;/h3H;1H3
InChI key
SOIFLUNRINLCBN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1
supp_hazards
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service