Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30736

Sigma-Aldrich

Sulfuric acid, fuming

puriss. p.a., 20% SO3

Synonym(s):

Oleum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4 · (SO3)x
CAS Number:
Molecular Weight:
98.08
Beilstein/REAXYS Number:
2037554
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)

grade

puriss. p.a.

assay

≤87% total SO3 basis

concentration

20-30% (free SO3)

impurities

≤0.0002% ammonium (NH4)
≤0.0005% heavy metals (as Pb)

ign. residue

≤0.01%

density

1.90 g/mL at 20 °C
1.925 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg

cation traces

As: ≤0.03 mg/kg
Fe: ≤50 mg/kg

SMILES string

O=S(=O)=O.OS(O)(=O)=O

InChI

1S/H2O4S.O3S/c1-5(2,3)4;1-4(2)3/h(H2,1,2,3,4);

InChI key

HIFJUMGIHIZEPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service