Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30530

Sigma-Aldrich

Sodium hydroxide solution

≥20%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

vapor pressure

3 mmHg ( 37 °C)

concentration

≥20%

impurities

≤0.002% heavy metals (as Pb)

anion traces

carbonate (as Na2CO3): ≤2%
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Fe: ≤10 mg/kg

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydroxide is an inorganic base. It participates in the oxidation of glycerol, catalyzed by Au/charcoal or Au/graphite.[1] It also participates in solvent-free aldol condensation reaction.[2]

Application

Sodium hydroxide may be employed for determination of specific surface area of colloidal silica, via titration.[3] It may be employed for direct oxidation of sodium borohydride, which was investigated by cyclic and linear voltammetry, chronoamperometry and chronopotentiometry.[4] It may be used to compose citrate buffer and borate buffers.[5]
It may be used in the synthesis of the following compounds:[6]
  • alkylaryl amidosulfobetaines
  • (3-[4-teroctylphenoxyethoxyethoxyethyl] dimethylammonio propane sulfonate (XOSB)
  • trisulfobetaines (TriSBn)

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Silvio Carrettin et al.
Chemical communications (Cambridge, England), (7)(7), 696-697 (2002-07-18)
Glycerol is oxidised to glyceric acid with 100% selectivity using either 1% Au/charcoal or 1% Au/graphite catalyst under mild reaction conditions (60 degrees C, 3 h, water as solvent).
Determination of specific surface area of colloidal silica by titration with sodium hydroxide.
Sears Jr GW.
Analytical Chemistry, 28(12), 1981-1983 (1956)
Chemoselective, solvent-free aldol condensation reaction.
Raston CL and Scott JL.
Green Chemistry, 2(2), 49-52 (2000)
M A Roberts et al.
Analytical chemistry, 69(11), 2035-2042 (1997-06-01)
This report describes a UV laser photoablation method for the production of miniaturized liquid-handling systems on polymer substrate chips. The fabrication of fluid channel and reservoir networks is accomplished by firing 200 mJ pulses from an UV excimer laser at
Kinetics of sodium borohydride direct oxidation and oxygen reduction in sodium hydroxide electrolyte: Part I. BH4- electro-oxidation on Au and Ag catalysts.
Chatenet M, et al.
Electrochimica Acta, 51(25), 5459-5467 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service