Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30414

Sigma-Aldrich

Sodium phosphate dibasic dodecahydrate

99-101% (calc. to the dried substance)

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 12H2O
CAS Number:
Molecular Weight:
358.14
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
assay:
99-101% (calc. to the dried substance)
form:
solid

assay

99-101% (calc. to the dried substance)

form

solid

impurities

Reducing matter, complies
≤0.001% Total nitrogen (N)
≤0.005% In water insoluble matters
≤0.5% NaH2PO4 (calc. to dried subst.)

pH

9.0-9.3 (20 °C, 5%)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤10 mg/kg
Pb: ≤2 mg/kg
Zn: ≤5 mg/kg

suitability

complies for appearance of solution

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.12H2O/c;;1-5(2,3)4;;;;;;;;;;;;/h;;(H3,1,2,3,4);12*1H2/q2*+1;;;;;;;;;;;;;/p-2

InChI key

DGLRDKLJZLEJCY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia Lin Chang et al.
Evidence-based complementary and alternative medicine : eCAM, 2012, 125247-125247 (2011-07-15)
The objectives of this study were to determine phytochemical compositions, chemiluminescence antioxidant activities, and neuroprotective effects on PC12 cells for water, methanol, and 95% ethanol extracts of the air-dried fruit of Terminalia chebula Retzius. The water extract afforded the greatest
Matthew H Crouthamel et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(11), 2625-2632 (2013-08-24)
Elevated serum phosphate has emerged as a major risk factor for vascular calcification. The sodium-dependent phosphate cotransporter, PiT-1, was previously shown to be required for phosphate-induced osteogenic differentiation and calcification of cultured human vascular smooth muscle cells (VSMCs), but its
Yuichiro Yano et al.
Journal of the American College of Cardiology, 65(4), 327-335 (2015-01-31)
Isolated systolic hypertension (ISH), defined as systolic blood pressure (SBP) ≥140 mm Hg and diastolic blood pressure (DBP) <90 mm Hg, in younger and middle-aged adults is increasing in prevalence. The aim of this study was to assess the risk
George G S Sandor et al.
International journal of cardiology, 179, 470-475 (2014-12-04)
Patients with Marfan (MFS) and Loeys-Dietz (LDS) syndromes have been shown to have abnormal aortic biophysical properties. The purpose of this study was to compare the effects of 12-months of therapy with atenolol or losartan on vascular function in young
Luciano Candilio et al.
Heart (British Cardiac Society), 101(3), 185-192 (2014-09-26)
Remote ischaemic preconditioning (RIPC), using brief cycles of limb ischaemia/reperfusion, is a non-invasive, low-cost intervention that may reduce perioperative myocardial injury (PMI) in patients undergoing cardiac surgery. We investigated whether RIPC can also improve short-term clinical outcomes. One hundred and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service