Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

256390

Sigma-Aldrich

Diethylene glycol dimethyl ether

spectrophotometric grade, ≥99%

Synonym(s):

2-Methoxyethyl ether, Bis(2-methoxyethyl) ether, Dimethyldiglycol, ‘Diglyme’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OCH2CH2)2O
CAS Number:
Molecular Weight:
134.17
Beilstein/REAXYS Number:
1736101
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

grade

spectrophotometric grade

vapor density

4.6 (vs air)

vapor pressure

3 mmHg ( 20 °C)

assay

≥99%

form

liquid

autoignition temp.

370 °F

expl. lim.

17.4 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

<0.050% water

evapn. residue

<0.001%

refractive index

n20/D 1.408 (lit.)

pH

7 (20 °C)

bp

162 °C (lit.)

mp

−64 °C (lit.)

density

0.944 g/mL at 20 °C (lit.)
0.939 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 225 nm Amax: 1.00
λ: 240 nm Amax: 0.50
λ: 260 nm Amax: 0.20
λ: 280 nm Amax: 0.08
λ: 320-400 nm Amax: 0.01

SMILES string

COCCOCCOC

InChI

1S/C6H14O3/c1-7-3-5-9-6-4-8-2/h3-6H2,1-2H3

InChI key

SBZXBUIDTXKZTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameHealth hazard

signalword

Danger

Hazard Classifications

Flam. Liq. 3 - Repr. 1B

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

134.6 °F - closed cup

flash_point_c

57 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Densities, refractive indices, speeds of sound, and shear viscosities of diethylene glycol dimethyl ether with ethyl acetate, methyl benzoate, ethyl benzoate, and diethyl succinate in the temperature range from 298.15 to 318.15 K.
Aminabhavi TM, et al.
Journal of Chemical and Engineering Data, 39(2), 251-260 (1994)
Eva Engel et al.
Analytical chemistry, 78(18), 6440-6447 (2006-09-15)
Tattooing is a widespread process of puncturing pigments into skin, whereas the resulting concentration inside the skin remains unknown. Many tattoo colorants are organic pigments, such as azo pigments, manufactured for other uses. To remove tattoos from skin, laser pulses
Qian Cheng et al.
Biomaterials, 30(25), 4203-4210 (2009-05-30)
Polyethylene glycol (PEG)-like films were grafted onto various substrates by plasma polymerization of diethylene glycol dimethyl ether (diglyme). Low-power diglyme plasma conditions yielded PEG-like films exhibiting nonfouling behavior, as evidenced by the significant decrease in protein adsorption and cell attachment
Kikuko Hayamizu et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 167(2), 328-333 (2004-03-26)
Pulsed gradient spin-echo (PGSE) NMR measurements of the self-diffusion coefficients of low viscosity liquids are greatly hampered by the effects of convection especially away from ambient temperature. Here we report on a new NMR tube designed to minimize the deleterious
Louise E Cullen et al.
Tree physiology, 25(5), 563-569 (2005-03-03)
The Jayme-Wise and diglyme-HCl methods for extracting cellulose from plant material for stable-isotope analysis differ considerably in ease of use, with the latter requiring significantly less time and specialized equipment. However, the diglyme-HCl method leaves a small lignin residue in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service