Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

246972

Sigma-Aldrich

Magnesium sulfate

anhydrous, ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgSO4
CAS Number:
Molecular Weight:
120.37
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

anhydrous

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

ReagentPlus®

assay

99%

SMILES string

[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

Inchi Key

CSNNHWWHGAXBCP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lex W Doyle
Current opinion in pediatrics, 24(2), 154-159 (2012-01-10)
Antenatal magnesium sulfate may reduce the excessive rates of cerebral palsy in survivors of very preterm birth. There are five randomized controlled trials of magnesium sulfate therapy given to the mother prior to very preterm birth which have reported neurological
Claire L O'Brien et al.
Gut, 63(10), 1596-1606 (2014-01-17)
Our aim was to determine whether or not specific microorganisms were transported selectively to lymph nodes in Crohn's disease (CD) by comparing node and mucosal microbial communities in patients and controls. We also sought evidence of dysbiosis and bacterial translocation.
Lex W Doyle et al.
Obstetrics and gynecology, 113(6), 1327-1333 (2009-05-23)
To systematically review rates of neurologic outcomes reported in childhood for the preterm fetus exposed to antenatal magnesium sulfate. We searched the Cochrane Pregnancy and Childbirth Group's Trials Register, CENTRAL (The Cochrane Library 2008, Issue 3), relevant references from retrieved
A P Morrison et al.
British journal of anaesthesia, 110(5), 702-712 (2013-03-28)
Spinal anaesthesia is the primary anaesthetic technique for many types of surgery. Adjuncts to the local anaesthetics (LA) used in spinal anaesthesia can exhibit undesirable side-effects, limiting their use, but magnesium may have advantages in this respect. We sought randomized
Kristin Luehders et al.
Development (Cambridge, England), 142(19), 3351-3361 (2015-10-08)
Small leucine-rich repeat proteoglycan (SLRP) family proteins play important roles in a number of biological events. Here, we demonstrate that the SLRP family member Asporin (ASPN) plays a crucial role in the early stages of eye development in Xenopus embryos.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service