Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24533

Sigma-Aldrich

Petrol

puriss., bp 90-100 °C

Synonym(s):

Gasoline, Petroleum benzin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
15101506
bp:
90-100 °C

grade

puriss.

impurities

≤0.005% S-compounds (as S)
≤0.005% non-volatile matter
≤0.01% water (Karl Fischer)
≤0.02% aromatic substances (as C6H5CH3)

refractive index

n20/D 1.395-1.400

bp

90-100 °C

density

0.705-0.730 g/mL at 20 °C

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Pertains only in Germany: unversteuert

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-56.2 °F

flash_point_c

-49 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Improved Gas Chromatographic Method for the Determination of C6-C12 Petroleum Hydrocarbons in Soil Water.
Voyevoda M, et al.
Clean: Soil, Air, Water, 36(2, 164-170 (2008)
GC methods for quantitative determination of benzene in gasoline.
Pavlova A and Ivanova R.
Acta Chromatographica , 215-225 (2003)
Hosein Karim et al.
Journal of injury & violence research, 7(2), 59-63 (2015-06-18)
To investigate factors related to prevention of self-immolation in west of Iran. In a case-control study, 30 consecutive cases of deliberate self-inflicted burns admitted to the regional burn centre (Imam Khomeini hospital in Kermanshah province, Iran) were compared with controls
Catherine F Poh et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(22), 6716-6722 (2006-11-24)
Genetically altered cells could become widespread across the epithelium of patients with oral cancer, often in clinically and histologically normal tissue, and contribute to recurrent disease. Molecular approaches have begun to yield information on cancer/risk fields; tissue optics could further
The use of a surgical glove in the prevention of tourniquet-associated chemical burns.
P J Tomlinson et al.
Annals of the Royal College of Surgeons of England, 90(3), 255-255 (2008-04-26)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service