Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24263

Sigma-Aldrich

Chloroform

technical, amylene stabilized, ≥99% (GC)

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein/REAXYS Number:
1731042
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

grade

technical

assay

≥99% (GC)

form

liquid

quality

amylene stabilized

refractive index

n20/D 1.444-1.446
n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Indications and contraindications for clinical psychotherapy].
A Marković
Medicinski arhiv, 33(1), 87-90 (1979-01-01)
Jameel A Feshitan et al.
Journal of colloid and interface science, 329(2), 316-324 (2008-10-28)
Microbubbles used as contrast agents for ultrasound imaging, vectors for targeted drug delivery and vehicles for metabolic gas transport require better size control for improved performance. Mechanical agitation is the only method currently available to produce microbubbles in sufficient yields
Z V Leonenko et al.
Biochimica et biophysica acta, 1509(1-2), 131-147 (2000-12-19)
We have used magnetic alternating current mode atomic force microscopy (MAC-AFM) to investigate the formation of supported phospholipid bilayers (SPB) by the method of vesicle fusion. The systems studied were dioleoylphosphatidylcholine (DOPC) on mica and mica modified with 3-aminopropyl-triethoxy-silane (APTES)
G F Ames et al.
Journal of bacteriology, 160(3), 1181-1183 (1984-12-01)
We introduce a method by which periplasmic proteins can be released rapidly, simply, and quantitatively by treating cells with chloroform. All the amino acid-binding proteins tested maintained their activity during chloroform treatment. This method makes practical the analysis of the
Gurman Singh Pall et al.
Nucleic acids research, 35(8), e60-e60 (2007-04-05)
The northern blot, or RNA gel blot, is a widely used method for the discovery, validation and expression analysis of small regulatory RNA such as small interfering RNA (siRNA), microRNA (miRNA) and piwi-interacting RNA (piRNA). Although it is straightforward and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service