Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

237140

Sigma-Aldrich

Lead(IV) oxide

ACS reagent, ≥97.0%

Synonym(s):

Lead (su)peroxide, Lead dioxide, Lead peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbO2
CAS Number:
Molecular Weight:
239.20
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
grade:
ACS reagent
assay:
≥97.0%

grade

ACS reagent

assay

≥97.0%

form

powder

reaction suitability

reagent type: catalyst
core: lead

dilution

(for analytical testing)

impurities

≤0.04% C compounds
≤0.2% acid insolubles

anion traces

chloride (Cl-): ≤0.002%
nitrate (NO3-): ≤0.02%
sulfate (SO42-): ≤0.05%

cation traces

Ca: ≤0.02%
Cu: ≤0.05%
K: ≤0.05%
Mn: ≤5 ppm
Na: ≤0.1%

SMILES string

O=[Pb]=O

InChI

1S/2O.Pb

InChI key

YADSGOSSYOOKMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Ox. Sol. 3 - Repr. 1A - STOT RE 2

target_organs

Blood,Central nervous system,Immune system,Kidney

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biljana Sljukić et al.
Analytica chimica acta, 587(2), 240-246 (2007-03-28)
Carbon powder modified with lead(IV) oxide was synthesised by a wet impregnation procedure from concentrated lead(II) nitrate solution. The impregnated powder was subsequently thermally treated at 823 K. Electrochemical and spectroscopic characterisation of the modified powder showed that lead(IV) oxide
L H Marcolino-Júnior et al.
Journal of pharmaceutical and biomedical analysis, 25(3-4), 393-398 (2001-05-30)
A flow injection spectrophotometric procedure was developed for determining levodopa in tablets. The determination of this drug was carried out by reacting it with lead(IV) dioxide immobilized in polyester resin packed in a solid-phase reactor and the dopachrome yielded was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service