Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

230421

Sigma-Aldrich

Mercury(II) nitrate monohydrate

ACS reagent, ≥98.0%

Synonym(s):

Mercuric nitrate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg(NO3)2 · H2O
CAS Number:
Molecular Weight:
342.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
assay:
≥98.0%
grade:
ACS reagent
form:
powder or crystals

grade

ACS reagent

assay

≥98.0%

form

powder or crystals

reaction suitability

reagent type: catalyst
core: mercury

reduction residue

≤0.01%

mp

79 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.002%

cation traces

Fe: ≤0.001%

SMILES string

O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Hg.2NO3.H2O/c;2*2-1(3)4;/h;;;1H2/q+2;2*-1;

InChI key

KVICROHOONHSRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Increasing interest in solid state NMR of heavy-metal nuclei results in the preparation of several Hg-thiocyanate and selenocyanate compounds as potential models.[1]

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Factors affecting Hg (II) adsorption on hybrid nanostructured silicas: influence of the synthesis conditions.
Perez-Quintanilla D and Sierra I.
Journal of Porous Materials, 21(1), 71-80 (2014)
Bowmaker, G.A. et al.
Inorganic Chemistry, 37, 1734-1734 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service