Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22153

Sigma-Aldrich

Celite® 560

filter aid, treated with sodium carbonate, flux calcined

Synonym(s):

Diatomaceous earth, flux-calcined

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:
NACRES:
SB.54

form

powder

quality

flux calcined
treated with sodium carbonate

composition

Al2O3, 1.1%
CaO, 6.54%
Fe2O3, 0.39%
Na2O + K2O, 1.28%
SiO2, 89.5%

loss

~0.2% loss on ignition, 900 °C

particle size

≤148.5 μm (56% (acc. to Cilas))

pH

>8.5 (25 °C, 10% in aq. suspension)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diatomaceous earth is made up of natural amorphous silica formed from the fossilized skeletons of diatoms.[1] It is extracted, crushed, dried and calcined with flux material. DE is widely used as filler in construction materials, paints and as an anti-caking agent for agricultural chemicals.[2] It has high water and oil absorption ability.[1]

Application

Diatomaceous earth may be used as an adsorbent for column chromatography.[3]

Caution

Not Intended for cGMP Parenteral Pharmaceutical Manufacturing

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G D Pegrum et al.
Journal of clinical pathology, 20(1), 65-69 (1967-01-01)
Methods of platelet adhesiveness to glass and celite have been developed. The technical merits of these methods have been compared with a method using adenosine diphosphate (ADP). The aspects of platelet stickiness measured by the various techniques may be somewhat
Enzymic interesterification of fats: factors influencing the choice of support for immobilized lipase.
Wisdom RA, et al.
Enzyme and Microbial Technology, 6(10), 443-446 (1984)
Induction of p53-independent growth inhibition in lung carcinoma cell A549 by gypenosides.
Journal of cellular and molecular medicine (2015).
Liu JS, et al.
Journal of Cellular and Molecular Medicine, 19(7), 1697-1709 (2015)
H Checkoway et al.
British journal of industrial medicine, 50(7), 586-597 (1993-07-01)
A cohort mortality study was conducted among workers from two plants in the diatomaceous earth mining and processing industry in California. Diatomaceous earth consists of the skeletal remains of diatoms. Exposure to amorphous (non-crystalline) and crystalline silica in the form
Manipulating surface wettability and oil absorbency of diatomite depending on processing and ambient conditions.
Ozen I, et al.
Applied Surface Science, 332, 22-31 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service